missing translation for 'onlineSavingsMsg'
Learn More

Iridium Standard: Ir @ 10 μg/mL in 2% HCl, VHG™

Product Code. 17943896
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943896 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17943896 Supplier VHG Labs Supplier No. VHGLIRH100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Iridium Standard: Ir @ 10 μg/mL in 2% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iridium
CAS 7439-88-5
Methods ICP/ICP-MS
Analyte or Component Names Iridium
Shipping Condition Room Temperature
Molecular Formula Ir
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym Ir
InChI Formula InChI=1S/Ir
SMILES [Ir]
IUPAC Name iridium
Solvent or Matrix Dilute HCl
Molecular Weight (g/mol) 192.217
Quantity 100 mL
Formula Weight 192.963 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.