missing translation for 'onlineSavingsMsg'
Learn More

Iridium Internal Standard: Ir @ 10 μg/mL in 2% HCl, VHG™

Product Code. 17972496
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972496 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17972496 Supplier VHG Labs Supplier No. VHGLISAIR100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Iridium Internal Standard: Ir @ 10 μg/mL in 2% HCl
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Iridium
CAS 7439-88-5
Methods ICP-MS
Analyte or Component Names Iridium
Shipping Condition Room Temperature
Molecular Formula Ir
Certifications/Compliance ISO 17034
Synonym Ir
InChI Formula InChI=1S/Ir
SMILES [Ir]
IUPAC Name iridium
Solvent or Matrix Dilute HCl
Molecular Weight (g/mol) 192.217
Quantity 100 mL
Formula Weight 192.963 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.