missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The IRF3 antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human IRF3. This antibody recognizes human, mouse, zebrafish antigen. The IRF3 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.
Specifications
Specifications
| Antigen | IRF3 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Recombinant Monoclonal |
| Clone | 2H6 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Peptide |
| Purification Method | Protein A purification |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?