missing translation for 'onlineSavingsMsg'
Learn More

IRAK4-IN-1, MedChemExpress

Product Code. 16496367
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496367 5 mg 5mg
16473638 10 mg 10mg
16462806 25 mg 25mg
16434726 100 mg 100mg
16410037 50 mg 50mg
16400949 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16496367 Supplier Medchem Express Supplier No. HY101922

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IRAK4-IN-1 is an interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor with an IC50 of 7 nM.

IRAK4-IN-1 is an interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor with an IC50 of 7 nM.

Specifications

Chemical Name or Material IRAK4-IN-1
Physical Form Solid
CAS 1820787-94-7
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H23N5O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 14.29 mg/mL (42.35 mM; Need ultrasonic)
SMILES N#CC1=CC2=C(N=CN=C2N[C@@H]3CC[C@@H](N4CCOCC4)CC3)C=C1
Molecular Weight (g/mol) 337.42
Formula Weight 337.42
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.