missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4 Antibody (SR1922), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00 EUR - 472.00 EUR
Specifications
| Antigen | IRAK4 |
|---|---|
| Clone | SR1922 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610214
|
Novus Biologicals
NBP3-21753-100ul |
100 μg |
472.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672625
|
Novus Biologicals
NBP3-21753-50ul |
50 μg |
349.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRAK4 Monoclonal antibody specifically detects IRAK4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| IRAK4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
| A synthesized peptide derived from human IRAK4 (Uniprot #: Q9NWZ3) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1922 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 51135 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title