Learn More
Memmert™ IQ/OQ Document with Test Data for IF75plus
Product Code. 17547305 Shop All Memmert Products
Change view
Click to view available options
:
For Use With: IF75plus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17547305 For Use With: IF75plus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17547305 Supplier Memmert™ Supplier No. IF75plusD00127
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
For Use With (Equipment) IF75plus Incubator
Product Type IQ/OQ Documentation
Includes Test Data