Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP410eco
HPP750eco
IPP1060eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- 27 measuring points (sizes 110 - 1060, 2200)
- 26 measuring points (size 1400)
- PQ checklist as support for validation by customer
Specifications
Specifications
| For Use With (Equipment) | IPP1060eco |
| Product Type | IQ/OQ Document with Device-specific Works Test Data |