Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature
Product Code. 17110155 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP410eco
HPP750eco
IPP1060eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17110155
Brand: Memmert™ IPP2200ecoD00127
For one free-selectable temperature value, Includestemperature distribution survey at Memmert to DIN 12880:2007-05
  • 27 measuring points (sizes 110 - 1060, 2200)
  • 26 measuring points (size 1400)
  • PQ checklist as support for validation by customer
Specifications
For Use With (Equipment) IPP2200eco
Product Type IQ/OQ Document with Device-specific Works Test Data