Learn More
Memmert™ IQ Document With Device-Specific Works Test Data
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP260eco
HPP410eco
HPP750eco
ICO105
ICO150
ICO240
ICO50
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Accessory
- OQ/ PQ check list as support for validation by customer
Specifications
Specifications
| For Use With (Equipment) | HPP1060eco |
| Product Type | IQ Document with Device-specific Works Test Data |