Learn More
Cytiva IPG Buffers
Shop All Cytiva Products
Click to view available options
pH Range:
3 to 10
3 to 10 nonlinear
3 to 11 nonlinear
4 to 7
6 to 11
Unit Size:
1mL
Each
Description
- Eliminates background staining
- Use with Immobiline DryStrip gels to improve protein solubility
- Ensures optimal conditions for IEF separation
- High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels
Order Info
Supplied in 1mL aliquots of water; order Immobiline Drystrip gels separately.
Specifications
Specifications
| Color | Colorless to Light Yellow |
| For Use With (Equipment) | Immobiline DryStrip Gels |
| pH Range | 3 to 11 nonlinear |
| Product Type | IPG Buffer |
| Form | Liquid |
| For Use With (Application) | IEF, 2-D Electrophoresis |
| Quantity | 1mL |
| Storage Requirements | 2° to 8°C |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu