missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP501 + Common metals, 100ppm: Al, Ca, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, V, Zn @ 100 μg/mL in 5% HNO3/tr HF., VHG™
Description
IP501 + Common metals, 100ppm: Al, Ca, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, V, Zn @ 100 μg/mL in 5% HNO3/tr HF.
Specifications
Specifications
| Chemical Name or Material | Aluminium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Nickel/Phosphorus/Potassium/Silicon/Sodium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-70-2, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1 |
| Methods | ICP |
| Analyte or Component Names | Aluminium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Nickel/Phosphorus/Potassium/Silicon/Sodium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3/HF |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 100 μg/mL/Calcium 100 μg/mL/Chromium 100 μg/mL/Copper 100 μg/mL/Iron 100 μg/mL/Lead 100 μg/mL/Magnesium 100 μg/mL/Manganese 100 μg/mL/Nickel 100 μg/mL/Phosphorus 100 μg/mL/Potassium 100 μg/mL/Silicon 100 μg/mL/Sodium 100 μg/mL/Vanadium 100 μg/mL/Zinc 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?