missing translation for 'onlineSavingsMsg'
Learn More

Iopamidol, MedChemExpress

Product Code. p-200096370
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16472196

Brand: Medchem Express HYB0684500mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) for a wide variety of diagnostic applications. Iopamidol contains amide and hydroxyl functionalities that can be exploited for the generation of the chemical exchange saturation transfer (CEST) contrast.

Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) for a wide variety of diagnostic applications. Iopamidol contains amide and hydroxyl functionalities that can be exploited for the generation of the chemical exchange saturation transfer (CEST) contrast.

Specifications

Chemical Name or Material Iopamidol
CAS 60166-93-0
Color White
Purity Grade Notes Research
Molecular Formula C17H22I3N3O8
Quantity 500 mg
Synonym B-15000 SQ-13396
Solubility Information H2O : 100 mg/mL (128.69 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (64.34 mM; Need ultrasonic)
SMILES O=C(C1=C(I)C(NC([C@@H](O)C)=O)=C(I)C(C(NC(CO)CO)=O)=C1I)NC(CO)CO
Molecular Weight (g/mol) 777.09
Formula Weight 777.09
Percent Purity 99.74%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.