Learn More
Thermo Fisher™ Genexus™ Dx cfTNA Purification Combo Kit
Shop All Thermo Scientific ProductsDescription
The Ion Torrent Genexus Dx Cell-Free Total Nucleic Acid (cfTNA) Purification Kit enables automated extraction and quantitation of TNA from cell-free plasma isolated from whole blood. This kit supports four runs of up to six samples each, utilizing our proven MagMAX technology and Genexus Dx Purification System for extraction and the onboard Qubit Fluorometer for post-extraction DNA quantitation.
The kit is intended for professional users, such as technicians who are trained in molecular biological techniques.
Benefits of the Genexus Dx cfTNA Purification Kit include:
- Consistent extraction results with MagMAX technology
- Reduced hands-on setup time using pre-filled consumables
- Maximized efficiency with automated Qubit quantitation
- Rapid turnaround times of as little as 3.25 hours from lysate to quantified nucleic acid
- Fast, automated procedure that allows 24 samples to be processed
The Genexus Dx cfTNA Purification Kit comes with all the reagents necessary to take a cfTNA sample from extraction to results. The kit includes the following components to provide a complete nucleic acid purification and quantification NGS workflow solution:
- Genexus Dx Cell-Free Total Nucleic Acid Purification Kit
- Genexus Dx Nucleic Acid Quantitation Kit
- Genexus Dx Purification Supplies 1 Kit
The kit contains plates prefilled with most of the reagents necessary to perform the extraction, requiring only a few pipetting steps per sample to set up the run. Simply remove the foil seal from the plate, add binding buffer, enzymes, and sample, then place the plate onto the Genexus Dx Purification System with onboard Qubit Fluorometer. After extraction, all nucleic acids are transferred directly to an archive plate and kept cool for up to one hour after run completion. The 48-well nucleic acid archive plate is barcoded to facilitate tracking of sample extractions across several stored archive plates.
Intended purpose
The Genexus Dx Cell-Free Total Nucleic Acid Purification Kit provides components for automated nucleic acid isolation, and quantification from plasma. The kit uses MagMAX magnetic bead-based for isolation and purification of cell-free total nucleic acid (cfTNA) from plasma. The isolated cfTNA is quantified using the onboard Qubit Fluorometer and fluorescence-based quantification reagents provided in the kit.
The product is intended for professional users, such as technicians who are trained in molecular biological techniques. The Genexus Dx Cell-Free Total Nucleic Acid Purification Kit is for in vitro diagnostic use.
CE-IVD according to IVDR. Not available in all countries.
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| Product Type | Purification FFPE samples - IVD |
| Quantity | 48 Samples |
For In Vitro Diagnostic Use.