Learn More
SI Analytics™ Ion-selective Indicator Electrode
Shop All SI Analytics Products
Click to view available options
Electrode:
Calcium electrode
Unit Size:
Each
Description
- Calcium electrode
- Fixed cable
- DIN plug
Specifications
Specifications
| Operating Range (Metric) | 0.02 to 40,000 mg/L |
| pH Range | 2.5 to 11 |
| Electrode | Calcium electrode |
| Temperature (Metric) Range | 0°C to 40°C |
| Includes | 1 m fixed cable with DIN plug |