missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Ion Chromatography (IC) Acrodisc™ Syringe Filters
These syringe filters are specifically designed for ion chromatography applications and optimized to provide the most consistent results when analyzing ionic species.
Brand: Cytiva (formerly Pall Lab) 4485
Additional Details : Weight : 0.47500kg
Description
- For research use only
Specifications
Supor (PES) | |
Non-sterile | |
Polypropylene | |
100/Pk. |
0.45 μm | |
1 cm2 | |
300/Cs. | |
13 mm |