missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Ion Chromatography (IC) Acrodisc™ Syringe Filters

These syringe filters are specifically designed for ion chromatography applications and optimized to provide the most consistent results when analyzing ionic species.

Brand:  Cytiva (formerly Pall Lab) 4483

248.90 EUR valid until 2024-05-06
Use promo code "21711" to get your promotional price.



Additional Details : Weight : 0.46550kg

 View more versions of this product

Product Code. 17174321

  • 262.00 EUR / Pack of 100
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

  • For research use only
Specifications

Specifications

Supor (PES)
Non-sterile
Polypropylene
100/Pk.
0.2 μm
3.9 cm2
300/Cs.
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers