Learn More
Cytiva (formerly Pall Lab) Ion Chromatography (IC) Acrodisc™ Syringe Filters
These syringe filters are specifically designed for ion chromatography applications and optimized to provide the most consistent results when analyzing ionic species.
Brand:  Cytiva (formerly Pall Lab) 4685
Additional Details : Weight : 0.94050kg
 View more versions of this product
Product Code. 17154391
  • 2275.00 EUR / Pack of 1000
Estimated Shipment: 01-07-2024
Description
  • For research use only
Specifications
Specifications
Supor (PES)
Non-sterile
Polypropylene
13 mm
0.45 μm
1 cm2
1000/Pk.