missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Ion Chromatography (IC) Acrodisc™ Syringe Filters

These syringe filters are specifically designed for ion chromatography applications and optimized to provide the most consistent results when analyzing ionic species.

Brand:  Cytiva (formerly Pall Lab) 4683

1851.07 EUR valid until 2024-05-06
Use promo code "21711" to get your promotional price.



Additional Details : Weight : 0.95090kg

 View more versions of this product

Product Code. 17144391

  • 2065.00 EUR / Pack of 1000
Estimated Shipment: 30-07-2024
to see stock.
Add to basket

Description

Description

  • For research use only
Specifications

Specifications

Supor (PES)
Non-sterile
Polypropylene
13 mm
0.2 μm
3.9 cm2
1000/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers