missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Ion Chromatography (IC) Acrodisc™ Syringe Filters

These syringe filters are specifically designed for ion chromatography applications and optimized to provide the most consistent results when analyzing ionic species.

Brand:  Cytiva (formerly Pall Lab) 4585

181.45 EUR valid until 2024-05-06
Use promo code "21711" to get your promotional price.



Additional Details : Weight : 0.58710kg

 View more versions of this product

Product Code. 17144361

  • 191.00 EUR / Pack of 50
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

  • For research use only
Specifications

Specifications

Supor (PES)
Non-sterile
Polypropylene
50/Pk.
0.45 μm
3.9 cm2
200/Cs.
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers