Learn More
Cytiva (formerly Pall Lab) Ion Chromatography (IC) Acrodisc™ Syringe Filters
These syringe filters are specifically designed for ion chromatography applications and optimized to provide the most consistent results when analyzing ionic species.
Brand: Cytiva (formerly Pall Lab) 4583
176.45 EUR valid until 2024-05-06
Use promo code "21711" to get your promotional price.
Additional Details : Weight : 1.23500kg
Description
- For research use only
Specifications
Supor (PES) | |
Non-sterile | |
Polypropylene | |
50/Pk. |
0.2 μm | |
3.9 cm2 | |
200/Cs. | |
25 mm |