missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodosulfuron-methyl-sodium, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Iodosulfuron-methyl-sodium |
| CAS | 144550-36-7 |
| CAS Max % | ≤ 100% |
| Density | 1.76 g/cm3 |
| Color | Beige to Colorless |
| Melting Point | 152°C |
| Synonym | Methyl 4-iodo-2-[3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)ureidosulfonyl]benzoate sodium salt |
| Molecular Weight (g/mol) | 529.24 g/mol |
| Quantity | 100 mg |
| Grade | Herbicides |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction