Learn More
Invitrogen™ ΦX174 RF DNA/Hae III Fragments
Description
Includes
ΦX174 RF DNA/Hae III Fragments are supplied at 0.5μg/μL in 10mM Tris-HCl (pH 7.4), 5mM NaCl, 0.1mM EDTA
ΦX174 RF DNA/Hae III Fragments are prepared by digesting purified ΦX174 RF DNA to completion with Hae III. These bands can be visualized by ethidium bromide, SYBR™ Safe, or SYBR Gold staining. Compatible with agarose gels.
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Concentration | 0.5 μg/μL |
| Form | Liquid |
| Kit Contents | ΦX174 RF DNA/Hae III Fragments are supplied at 0.5μg/μL in 10mM Tris-HCl (pH 7.4), 5mM NaCl, 0.1mM EDTA |
| Quantity | 80 μL |
| Range | 72 to 1353 bp |
| Product Type | DNA Molecular Weight Marker |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.