Learn More
Invitrogen™ TrackIt™ λ DNA/Hind III Fragments
Description
Includes
500μL of TrackIt™ λ DNA/Hind III Fragments are supplied at a concentration of 0.1μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.4% Orange G, and 5% glycerol; For 100 applications
TrackIt™ λ DNA/Hind III Fragments are prepared by digesting λ DNA (cIind1ts857 Sam7) to completion with Hind III. Bands can be visualized by ethidium bromide, SYBR™ Safe, or SYBR Gold staining. The TrackIt DNA ladders are ready-to-load and minimize time spent thawing, diluting, and adding sample buffer to your DNA ladders. Simply transfer your TrackIt DNA ladder from the vial to the gel. TrackIt ladders are stable at room temperature and can be stored on your benchtop. TrackIt DNA ladders offer two tracking dyes, one that runs behind the sample and one that runs ahead of the sample. The tracking dyes serve as visual markers for migration during electrophoresis and indicate when maximum resolution of the DNA fragments has been achieved. Additionally, visualization of DNA bands is not obscured by the tracking dyes because they run outside the limits of most DNA samples.
- Ready-to-load convenience
- Better tracking with two dyes to indicate when maximum resolution has been achieved
- TrackIt dyes that run outside the limits of most DNA samples, preventing masking of sample DNA bands
- Compatible with agarose and E-Gel™ gels
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice
Specifications
Specifications
| Concentration | 0.1 μg/μL |
| Form | Liquid |
| Kit Contents | 500μL of TrackIt™ λ DNA/Hind III Fragments are supplied at a concentration of 0.1μg/μL in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.4% Orange G, and 5% glycerol |
| Quantity | 100 Applications |
| Range | 125 bp to 23.1kb |
| Product Type | DNA Molecular Weight Marker |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.