Learn More
Invitrogen™ PureLink™ HiPure BAC Buffer Kit
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Description
Includes
The PureLink HiPure BAC Buffer Kit includes Resuspension Buffer (R3) (250mL), Lysis Buffer (L7) (250mL), Precipitation Buffer (N3) (250mL), and RNase A (20 μg/mL) (5mL). All components of the PureLink HiPure BAC Buffer Kit are shipped at room temperature. Store all components at room temperature upon receipt.
- PureLink HiPure BAC Buffer Kit provides additional buffers and reagents for performing BAC (Bacterial Artificial Chromosome) DNA isolation using PureLink HiPure Plasmid Kits
- PureLink HiPure Plasmid Kits utilize proprietary anion-exchange resin which allows you to purify high-quality BAC DNA from E. coli cultures
- BAC DNA requires the use of large bacterial culture volumes to obtain a reasonable DNA yield
- From 100 mL culture, expect to purify ∽40 μg of BAC DNA
- PureLink HiPure BAC Buffer Kit provides reagents sufficient for five BAC Maxipreps, or 25 BAC Midipreps, or 100 BAC Minipreps
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | The PureLink™ HiPure BAC Buffer Kit includes Resuspension Buffer (R3) (250 mL), Lysis Buffer (L7) (250 mL), Precipitation Buffer (N3) (250 mL), and RNase A (20 mg/mL) (5 mL). All components of the PureLink™ HiPure BAC Buffer Kit are shipped at room temperature. Store all components at room temperature upon receipt. |
| Format | Kit |
| Isolation Technology | Anion Exchange Resin |
| Sample Type | Bacterial Culture |
| Final Product Type | BAC DNA |
| For Use With (Application) | Sequencing, Cloning, Transformation, Transfection, In Vitro Transcription |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Product Line | PureLink |
| Product Type | BAC Buffer Kit |
| Quantity | 1 kit |
| Show More |