missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq PCRx DNA Polymerase

Product Code. 10554033
Click to view available options
:
500 reactions
This item is not returnable. View return policy

Product Code. 10554033

Brand: Invitrogen™ 11509015

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Platinum Taq PCRx DNA Polymerase is supplied with 10X PCR buffer, 50mM MgCl2,50mM MgSO4, 10X PCRx Enhancer Solution, and 10X PCRx Amplification Buffer.

Especially effective for problematic templates such as GC-rich DNA

Platinum Taq PCRX DNA Polymerase is Platinum Taq DNA Polymerase supplied with the PCRX Enhancer System for optimizing PCR of problematic and/or GC-rich templates.

  • Higher primer specificity, broader magnesium concentration, broader annealing temperature, and improved thermostabilty of Taq DNA polymerase
  • Amplification of PCR products up to 5kb in size
  • A system that is licensed and qualified for PCR
  • Applications: Amplification of DNA from complex genomic, viral, and plasmid templates and RT-PCR, especially effective for problematic templates such as GC-rich DNA
  • Unit Definition: One unit incorporates 10nmol of deoxyribonucleotide into DNA in 30 min at 74°C

GC-Rich PCR, Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR

Order Info

Shipping Condition: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Platinum Taq PCRx DNA Polymerase is supplied with 10X PCR buffer, 50mM MgCl2 ,50mM MgSO4, 10X PCRx Enhancer Solution, and 10X PCRx Amplification Buffer. Store at -20°C. Guaranteed stable for 6 months when properly stored.
GC-Rich PCR Performance High
Polymerase Platinum Taq DNA Polymerase
Exonuclease Activity 5' - 3'
Quantity 500 Reactions
Fidelity (vs. Taq) 1 X
No. of Reactions 500
Overhang 3'-A
Size (Final Product) 5 kb or less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.