Learn More
Invitrogen™ pcDNA™3.1/Zeo (-) Mammalian Expression Vector
Description
Includes
20μg of this pcDNA3.1/Zeo(-) vector, as well as an expression control, are supplied supercoiled and lyophilized.
- Contains Zeocin selectable marker and reverse-orientation multiple cloning site
- Cytomegalovirus (CMV) enhancer-promoter for high-level expression
- Large multiple cloning site in either forward (+) or reverse (-) orientations
- Bovine Growth Hormone (BGH) polyadenylation signal and transcription termination sequence for enhanced mRNA stability
- SV40 origin for episomal replication and simple vector rescue in cell lines expressing the large T antigen (i.e.,COS-1 and COS-7)
- Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Transfection |
| Promoter | CMV |
| Product Type | Mammalian Expression Vector |
| Selection Agent (Eukaryotic) | Zeocin™ |
| Content And Storage | 20 μg of this pcDNA™3.1/Zeo(-) vector, as well as an expression control, are supplied supercoiled and lyophilized. Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored. |
| Protein Tag | Untagged |
| Cloning Method | Restriction Enzyme/MCS |
| Quantity | 20 μg |
| Vector | pcDNA |
| Show More |