Learn More
Invitrogen™ Optifect™ Transfection Reagent
Description
Includes
One 1mL vial.
Optifect Reagent can also be used with cells that are sensitive to Lipofectamine™ 2000 or other commercially available transfection reagents. Optifect Reagent facilitates high levels of protein expression from sparsely plated cells. Useful in applications such as studies of cell surface proteins, post-transfection proliferation assays, or other situations where cells must be allowed to grow significantly following transfection. Optifect has been shown to be less toxic to cells compared to other commercially available transfection reagents.
Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Noncoding RNA Research
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Content And Storage | One unit vial contains of reagent. Store at +4°C. Guaranteed stable for 6 months when properly stored. |
| Cell Type | Established Cell Lines, Primary Cells, Hard-to-Transfect Cells |
| Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
| Sample Type | Plasmid DNA |
| Transfection Technique | Lipid-based Transfection |
| Vector | pLATE |
| For Use With (Application) | Transfection |
| High-throughput Compatibility | High-throughput Compatible |
| Product Line | Optifect |
| Product Type | Transfection Reagent |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.