missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Opener

Product Code. 10573293
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10573293 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10573293 Supplier Invitrogen™ Supplier No. G530001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A simple device specifically designed to quickly and efficiently open an E-Gel™ cassette

Simply place the E-Gel cassette into the E-Gel Opener and turn the knob to tighten. The E-Gel Opener uses two steel blades to safely and quickly pop open the E-Gel cassette without harming your gel. In only a few minutes and with minimal effort, your E-Gel will be ready for subsequent procedures.

  • The E-Gel Opener is safe and easy to use
  • Anodized aluminum construction for maximum durability
  • Allows purification of DNA fragments from the gel, transfer samples onto a membrane for Southern blot analysis, or post-stain Clear EGels

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) E-Gel™ Cassettes
Type E-Gel Opener
Quantity 1 Each

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.