missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Mouse CD4

Product Code. 10706283
Click to view available options
Quantity:
5 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10706283

Brand: Invitrogen™ 11445D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

5mL DynabeadsMouse CD4 (L3T4) coated with anti-L3T4 monoclonal antibody. The L3T4 antigen is expressed on mouse thymocytes and the T helper subsets of mature T cells of all common mouse strains

Dynabeads™ Mouse CD4 are magnetic beads that provide a robust and versatile tool to isolate or deplete mouse CD4+ T-cells directly from any sample.

Dynabeads™ Mouse CD4 are magnetic beads that provide a robust and versatile tool to isolate or deplete mouse CD4+ T-cells directly from any sample, including tissue digests such as spleen or lymph nodes, whole blood, bone marrow, and mononuclear cells (MNC).
• Rapid isolation of mouse CD4+ T-cells from any sample—no columns required
• Positive isolation for molecular downstream assays or efficient depletion of mouse CD4+ T cells

Excellent Yield, Purity, and Viability of Recovered Cells
Dynabeads™ Mouse CD4 are uniform, superparamagnetic beads (4.5 μm diameter) that enable easy isolation of mouse CD4+ cells from spleen or lymph nodes, although any single-cell suspension may be used. Dynabeads™ Mouse CD4 beads are coated with a primary monoclonal antibody specific for the CD4 membrane antigen mainly expressed on mouse T helper cells. Dynabeads™ Mouse CD4 binds to CD4+ cells during a short incubation period in a tube, after which the bead-bound cells are separated from unbound cells using a magnet. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity, recovery and viability of the isolated CD4+ cells.

Ideal for Both Depletion and Positive Isolation for Downstream Molecular Assays
Because of the powerful magnets used for separation, Dynabeads™ Mouse CD4 can be used in viscous samples such as single cell suspensions from tissue, whole blood and bone marrow, and enable efficient depletion of mouse CD4+ cells in about 30 minutes. Dynabeads™ Mouse CD4 will also afford excellent recovery of high-purity, viable cells when used for positive isolation for downstream studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate mouse CD4+ cells for cell-based applications, or you need to check your samples with flow cytometry, you should use DETACHaBEAD™ Mouse CD4 (Cat. No. 12406D) in combination with this products (for bead- and antibody-free cells) or Dynabeads™ FlowComp Mouse CD4 Cells (Cat. No. 11461D) (complete kit for bead-free cells).

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (CD4)
Content And Storage This Product Contains: 5 mL Dynabeads™Mouse CD4 (L3T4) coated with anti-L3T4 monoclonal antibody. The L3T4 antigen is expressed on mouse thymocytes and the T helper subsets of mature T cells of all common mouse strains.

Store at 2°C to 8°C.
Product Type Antibody Coated Bead
Isolation Technology Negative Isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Reactivity Mouse
Sample Type Lymph Node, Blood, Spleen
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 cells per isolation
Target Species Mouse
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.