Learn More
Invitrogen™ λ DNA/Hind III Fragments
Description
Includes
λDNA/Hind III Fragments are supplied at 0.5μg/μL in 10mM Tris-HCl (pH 7.4), 5mM NaCl, 0.1mM EDTA
λDNA/Hind III Fragments are prepared by digesting λ DNA (cIind1ts857 Sam7) to completion with Hind III. Bands can be visualized by ethidium bromide, SYBR™ Safe, or SYBR Gold staining. Compatible with agarose gels.
- Gel Compatibilitiy: Agarose Gels
- Size Range: 125bp to 23.1kb
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Approved for shipment on Wet or Dry Ice
Specifications
Specifications
| Concentration | 0.5 μg/μL |
| Form | Liquid |
| Kit Contents | λDNA/Hind III Fragments are supplied at 0.5μg/μL in 10mM Tris-HCl (pH 7.4), 5mM NaCl, 0.1mM EDTA |
| Quantity | 500 μg |
| Range | 125 bp to 23.1kb |
| Product Type | DNA Molecular Weight Marker |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.