missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 250 bp DNA Ladder

Product Code. 11588626
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
75µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11588626 50 μL 75µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11588626 Supplier Invitrogen™ Supplier No. 10596013

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Includes

250bp DNA Ladder is supplied at 1μg/μL in 10mM Tris-HCl (pH 7.5), 1mM EDTA Volume: 50μL.

Suitable for sizing double-stranded DNA from 250 bp to 3.5 kb

Prepared from a plasmid containing repeats of a 250-bp DNA fragment, the ladder consists of 14 blunt fragments ranging in length from 250 to 3,500bp at 250-bp increments. For easy reference on agarose gels, the 1,000bp band is two- to three-times brighter than the other bands in the ladder. This ladder can be stained with ethidium bromide or radiolabeled using T4 polynucleotide kinase or T4 DNA polymerase.

  • Less waste, sustainable packaging
  • Compatible with agarose gels

Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 μg/μL
Form Liquid
Kit Contents 250bp DNA Ladder is supplied at 1μg/μL in 10mM Tris-HCl (pH 7.5), 1mM EDTA
Quantity 50 μL
Range 0.25 to 3.5kb
Product Type DNA Molecular Weight Marker

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.