missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intron-Binding Protein Of 160 KDa Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 EUR - 470.00 EUR
Specifications
| Antigen | Intron-Binding Protein Of 160 KDa |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18326284
|
Novus Biologicals
NBP3-03714-20UL |
20 μL |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331764
|
Novus Biologicals
NBP3-03714-100UL |
100 μL |
470.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Intron-Binding Protein Of 160 KDa Polyclonal specifically detects Intron-Binding Protein Of 160 KDa in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Intron-Binding Protein Of 160 KDa | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 9716 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| AQR, aquarius homolog (mouse), FSAP164, Functional Spliceosome-Associated Protein 164, IBP160, Intron-Binding Protein Aquarius | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human Intron-Binding Protein Of 160 KDa (NP_055506.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title