Learn More
Integrin alpha 2b/CD41 Mouse anti-Human, Clone: 745201, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  R&D Systems MAB7616-SP
Encompass
Product Code. 15798734
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions