missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220 Antibody (83-7), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81039H
This item is not returnable.
View return policy
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Insulin R/CD220 | |
| Monoclonal | |
| HRP | |
| PBS | |
| INSR | |
| Epitope within amino acids 140-301 (the cysteine rich region) of the extracellular domain of the human Insulin R/CD220. | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3643 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 83-7 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognises the extracellular domain of the Insulin R/CD220. Amino acids 469 - 592 (exon 7 and 8) of Insulin R/CD220. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction