missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Insulin, Human Recombinant, MP Biomedicals™

Product Code. 11446301 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446301 10mg 10mg
11456301 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446301 Supplier MP Biomedicals™ Supplier No. 0219390010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Insulin is a primary polypeptide hormone in humans.

Insulin is responsible for controlling the cellular uptake, utilization and storage of glucose, amino acids and fatty acids while inhibiting the breakdown of glycogen, protein and fat. It is produced by pancreatic beta cells.

  • Two-chain polypeptide hormone produced by the β-cells of pancreatic islets.
  • Its molecular weight is ∼5800 Da.
  • The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond.
  • Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.
  • 1 unit corresponds to the efficiency of 0.04167mg international standard substance.
  • An HPLC method exists to test the potency.
  • Typical working concentration is 1 to 10μg/mL for cell culture.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C.
For Use With (Application) Methods for immobilizing insulin on polystyrene dishes have been reported for applications involving protein-free cell cultures.
Quantity 10mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.