Learn More
Bochem™ Instruments Tray
Click to view available options
Dimensions (L x W x H):
180 x 90 x 30 mm
250 x 150 x 60 mm
Unit Size:
Each
Description
- Stainless Steel
- Rounded edges
- Lid included
Specifications
Specifications
| Dimensions (L x W x H) | 180 x 90 x 30 mm |
| Height (Metric) | 30 mm |
| Length (Metric) | 180 mm |
| Material | Stainless Steel |
| Type | Instrument Tray |
| Width (Metric) | 90 mm |
| Includes | Lid |
| Shape | Rectangular |