Learn More
Cytiva Insert Release
Product Code. 17154721
Change view
Click to view available options
Volume (Metric):
20 to 60 mL (Sample)
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
17154721 20 to 60 mL (Sample) Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17154721 Supplier Cytiva Supplier No. FD003X65
Only null left
Add to basket
Add to basket
Insert release for use with protein purification and sterile filtration workflows.
  • Utilizes proprietary high-performance multi-layer filter membranes
  • Plates offer time savings strong performance claims and streamlined workflow improvements
  • Each plate comes with a V-shaped bottom collection plate and lid
TRUSTED_SUSTAINABILITY
Specifications
Description EFA: 15.2 cm2
For Use With (Equipment) Jumbosep Centrifugal Device
Material (Membrane) PES
Sterility Non-sterile
Housing Material Polypropylene
Quantity 24/Pk.
Volume (Metric) 20 to 60 mL (Sample)