Learn More
Miele Professional™ Insert
Product Code. 10560672 Shop All Miele Products
Change view
Click to view available options
:
Dimensions (D x W x H): 517 x 535 x 502mm
Product Code.
10560672 Dimensions (D x W x H): 517 x 535 x 502mm
1 options
Product Code. 10560672
Brand: Miele Professional™ 3809410
This item requires a price update prior to being ordered.
Request a Price for this item here.
Mobile unit E 340 for the optimum loading of narrow necked glassware
  • Half can be equipped with other inserts
  • Equipped with 19 injector nozzles
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Product Type Mobile Unit for Narrow-necked Glassware
Color Stainless Steel
Holds 19 Narrow-necked glassware
For Use With (Equipment) Washer-disinfectors, laboratory
Depth (Metric) 517 mm
Height (Metric) 502 mm
Width (Metric) 535 mm