Learn More
Miele Professional™ Insert
Product Code. 11646040 Shop All Miele Products
Click to view available options
:
Dimensions (D x W X H): 440 x 205 x 35mm
Product Code. 11646040
Brand: Miele Professional™ 12669920
This item requires a price update prior to being ordered.
Request a Price for this item here.
Insert E 494 for the gentle holding of 5 microtiter plates
For microtiter plates
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Color Stainless Steel
Holds 5 microtiter plates
For Use With (Equipment) Washer-disinfectors, laboratory
Depth (Metric) 440 mm
Height (Metric) 35 mm
Width (Metric) 205 mm