missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inorganic Pyrophosphatase/PPA1 Antibody (SR1552), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22572-100ul
This item is not returnable.
View return policy
Description
Inorganic Pyrophosphatase/PPA1 Monoclonal antibody specifically detects Inorganic Pyrophosphatase/PPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Inorganic Pyrophosphatase/PPA1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1552 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061 | |
| A synthesized peptide derived from human Inorganic Pyrophosphatase/PPA1 (Uniprot #: Q15181) | |
| 100 μg | |
| metabolism, Signal Transduction | |
| 5464 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction