Learn More
Eppendorf™ Innova™ 40 Incubator Shaker
Product Code. 11360015
Change view
Click to view available options
Orbit:
19 mm
25 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit unitSize
11360015 25 mm Each
12508886 19 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 11360015 Supplier Eppendorf™ Supplier No. M12990092
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Use these versatile incubator shakers for support high-speed applications and heavy workloads. New Brunswick Scientific™ Innova™ 40 Incubator Shakers provide very broad temperature capabilities for culturing a wide variety of organisms in flasks up to 3 liters.
  • Exceptionally quiet operation provides a favorable work environment
  • Triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble free operation for years
  • Compact benchtop model – fits easily on the bench and holds flasks up to 3L
  • Clear cover allows easy viewing of the cultures and lifts easily to access all samples
  • Versatile accessory platform (sold separately) accommodates flasks up to 3L as well as test tube racks and Microplate holders
  • Certified. Meets cETL, UL, CSA and CE standards
TRUSTED_SUSTAINABILITY
Specifications
Orbit 25 mm
Speed Range 25 to 500 rpm
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Holds 3 L
Product Type Incubator Shaker
Frequency 50 Hz
Depth (Metric) 76.2 cm
Height (Metric) 101.6 cm
Width (Metric) 55.9 cm
Model Innova 40
Timer Range 0.01 to 99:59 hr.