Learn More
Mettler Toledo™ InLab™ Sensor Cables: MultiPin™ Connection
Product Code. 15599641 Shop All Mettler Toledo Products
Change view
Click to view available options
Connector Types:
MultiPin-BNC/1 x 4mm banana
MultiPin-BNC/RCA (cinch)
MultiPin-BNC/RCA (cinch), IP67 waterproof
MultiPin-DIN 19262/1 x 4mm banana
MultiPin-DIN/RCA (cinch)
MultiPin-Lemo 00/2 x 4mm banana
Length (Metric) Cable:
1.2 m
1.8 m
3.0 m
5.0 m
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Length (Metric) Cable unitSize
15599641 MultiPin-BNC/RCA (cinch) 3.0 m Each
15529651 MultiPin-DIN/RCA (cinch) 1.2 m Each
15509651 MultiPin-BNC/RCA (cinch) 5.0 m Each
15519651 MultiPin-BNC/1 x 4mm banana 1.2 m Each
15559651 MultiPin-BNC/RCA (cinch), IP67 waterproof 1.8 m Each
15503181 MultiPin-BNC/RCA (cinch) 1.2 m Each
15539651 MultiPin-DIN 19262/1 x 4mm banana 1.2 m Each
15549651 MultiPin-Lemo 00/2 x 4mm banana 1.2 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15599641 Supplier Mettler Toledo™ Supplier No. 30281897
Only null left
Add to basket
Add to basket
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter with compatible connector.
  • Detachable
  • Resuable
TRUSTED_SUSTAINABILITY
Specifications
Connector Types MultiPin-BNC/RCA (cinch)
For Use With (Equipment) Mettler Toledo InLab Sensors
For Use With (Application) To connect Mettler Toledo InLab sensors to any meter
Length (English) Cable 9.8 ft.
Length (Metric) Cable 3.0 m