Learn More
B Braun™ Injekt™ Solo Disposable Centric Luer Two-Part Syringe
Product Code. 10354641
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
20 mL
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10354641 5 mL Pack of 100
12762637 2 mL Pack of 100
10292941 20 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10354641 Supplier B Braun™ Supplier No. 4606710V
Manufactured from polypropylene/polyethylene. B. Braun™ Injekt™ Solo Disposable Centric Luer-lock Two-Part Syringe has high contrast graduations for ideal readability.
  • Made of polypropylene/polyethylene
  • „Green plunger
  • „Highly transparent barrel
  • „High contrast black graduation, permanent marking, ideal readability
  • „Safe plunger backstop
  • „Easy aspiration up to maximum volume
  • „Latex-free/PVC-free
  • „Silicone oil-free
  • Single sterile-packed (in box of 100 pcs)
Specifications
Certifications/Compliance ISO 7886-1
Material Polyethylene, Polypropylene
Color Green Plunger
Description Black graduation
For Use With (Application) Ideal for simple injections, vaccinations and more
Latex Free Yes
Product Type Centric Luer-lock 2-part Syringe
Disposable Single-use
Quantity 100/Pk.
Volume (Metric) 5 mL
Connection Type Centric Luer Lock
Graduated Yes