missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inhibin beta A Rabbit anti-Human, Mouse, Rat, Clone: S03-1F0, Novus Biologicals™
Rabbit Monoclonal Antibody
286.00 EUR - 581.00 EUR
Specifications
| Antigen | Inhibin beta A |
|---|---|
| Clone | S03-1F0 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18324667
|
Novus Biologicals
NBP3-15047-25UL |
25 μg |
286.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331405
|
Novus Biologicals
NBP3-15047-100UL |
100 μg |
581.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inhibin beta A Monoclonal antibody specifically detects Inhibin beta A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Inhibin beta A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Activin beta-A chain, beta A (activin A, activin AB alpha polypeptide), EDF, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FRP, FSH-releasing protein, inhibin beta A chain, inhibin, beta A, Inhibin, beta-1 | |
| A synthetic peptide of human Inhibin beta A (Uniprot#: P08476) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S03-1F0 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 3624 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title