missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H3N2 HA1 (A/Aichi/2/1968) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581698
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin/HA1.
Specifications
| Influenza A H3N2 HA1 (A/Aichi/2/1968) | |
| Polyclonal | |
| Unconjugated | |
| flu hemagglutinin; influenza hemagglutinin | |
| Recombinant Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin/HA1 Protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction