missing translation for 'onlineSavingsMsg'
Learn More

INF39, MedChemExpress

Product Code. 16460017
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460017

Brand: Medchem Express HY101868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress INF39 is an irreversible and noncytotoxic NLRP3 inhibitor.

INF39 is an irreversible and noncytotoxic NLRP3 inhibitor.

Specifications

Chemical Name or Material INF39
CAS 866028-26-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C12H13ClO2
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (445.08 mM; Need ultrasonic)
SMILES O=C(OCC)C(CC1=CC=CC=C1Cl)=C
Molecular Weight (g/mol) 224.68
Formula Weight 224.68
Percent Purity 99.23%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.