Learn More
Indole-3-acetic acid Sodium Salt, TRC
Specifications
Specifications
| CAS | 6505-45-9 |
| Molecular Formula | C10H8NNaO2 |
| Quantity | 1 g |
| Synonym | Indole-3-acetic Acid Monosodium Salt, 3-Indoleacetic Acid Sodium Salt, Sodium (3-indolyl)Acetate, Sodium Indole-3-acetate, Sodium Indoleacetate, beta-IAA Sodium Salt |
| InChI Formula | InChI=1S/C10H9NO2.Na/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9;/h1-4,6,11H,5H2,(H,12,13);/q;+1/p-1 |
| SMILES | [Na+].[O-]C(=O)Cc1c[nH]c2ccccc12 |
| IUPAC Name | sodium;2-(1H-indol-3-yl)acetate |
| Molecular Weight (g/mol) | 197.17 |
| Formula Weight | 197.0453 |
| Percent Purity | >95% |
| Show More |