missing translation for 'onlineSavingsMsg'
Learn More

(-)-Indolactam V, MedChemExpress

Product Code. p-200098231
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457807 5 mg 5mg
16421239 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16457807 Supplier Medchem Express Supplier No. HY12307

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-Indolactam V is a PKC activator, with Kis of 3.36 nM, 1.03 μM for η-CRD2 (PKCη surrogate peptide), γ-CRD2 (PKCγ surrogate peptide), and Kds of 5.5 nM (η-C1B), 7.7 nM (ε-C1B), 8.3 nM (δ-C1B), 18.9 nM (β-C1A-long), 20.8 nM (α-C1A-long), 137 nM (β-C1B), 138 nM (γ-C1A), 213 nM (γ-C1B), and has antitumor activity.

(-)-Indolactam V is a PKC activator, with Kis of 3.36 nM, 1.03 μM for η-CRD2 (PKCη surrogate peptide), γ-CRD2 (PKCγ surrogate peptide), and Kds of 5.5 nM (η-C1B), 7.7 nM (ε-C1B), 8.3 nM (δ-C1B), 18.9 nM (β-C1A-long), 20.8 nM (α-C1A-long), 137 nM (β-C1B), 138 nM (γ-C1A), 213 nM (γ-C1B), and has antitumor activity.

Specifications

Chemical Name or Material (-)-Indolactam V
Physical Form Solid
CAS 90365-57-4
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H23N3O2
For Use With (Application) Cancer-Kinase/protease
Synonym Indolactam V
Solubility Information DMSO : 50 mg/mL (165.90 mM; Need ultrasonic)
SMILES CC(C)[C@@H](C1=O)N(C)C2=C3C(NC=C3C[C@H](N1)CO)=CC=C2
Molecular Weight (g/mol) 301.38
Formula Weight 301.38
Percent Purity 98.75%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.