Learn More
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2 with certificate
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
76.96 EUR
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
15535099
|
Fisherbrand™
|
5 mg |
76.96 EUR
1 unit |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15575099
|
Fisherbrand™
|
100 mg |
76.96 EUR
1 unit |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15565099
|
Fisherbrand™
|
50 mg |
76.96 EUR
1 unit |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15545099
|
Fisherbrand™
|
10 mg |
76.96 EUR
1 unit |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15585099
|
Fisherbrand™
|
200 mg |
76.96 EUR
1 unit |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15595099
|
Fisherbrand™
|
500 mg |
76.96 EUR
1 unit |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15555099
|
Fisherbrand™
|
20 mg |
76.96 EUR
1 unit |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15525099
|
Fisherbrand™
|
2 mg |
76.96 EUR
1 unit |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15515099
|
Fisherbrand™
|
1 mg |
76.96 EUR
1 unit |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
- Class E2 calibration weight
- In compliance with OIML R111
- Delivered with a calibration certificate guaranteeing traceability
Specifications
E2 | |
Stainless Steel |