Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1
Product Code. 15585149 Shop All Fisherbrand Products
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
Product Code. 15585149
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
  • Class M1 stainless steel calibration weight
  • In compliance with OIML R111
Specifications
Classification M1
Material Stainless Steel
Weight (Metric) 10 g
Mass 10 g
Shape Cylindrical